Product Details

SNP ID
rs61198573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:43635823 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTGGAGGCGCGGGCTTCCCATGA[G/T]GCCCCGCGAGACCTTTATTCTAACC
Phenotype
MIM: 607434 MIM: 612648
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GTPBP2 PubMed Links

Gene Details

Gene
GTPBP2
Gene Name
GTP binding protein 2
There are no transcripts associated with this gene.

Gene
MAD2L1BP
Gene Name
MAD2L1 binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001003690.1 Intron NP_001003690.1
NM_014628.2 Intron NP_055443.1
Gene
RSPH9
Gene Name
radial spoke head 9 homolog
There are no transcripts associated with this gene.

View Full Product Details