Product Details

SNP ID
rs115969600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:42932364 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCATGCTGAAAGCAGGGGCGGGACC[C/G]GGGCGGTCTTCCAGCAGGGAAAATG
Phenotype
MIM: 611839 MIM: 176842
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MRPL32 PubMed Links

Gene Details

Gene
MRPL32
Gene Name
mitochondrial ribosomal protein L32
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031903.2 Intron NP_114109.1
Gene
PSMA2
Gene Name
proteasome subunit alpha 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002787.4 Intron NP_002778.1

View Full Product Details