Product Details

SNP ID
hCV25594856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:235335214 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACACTGAAAGATGATTAGTTTCATG[G/T]GATCTTTATGTTTCTCCTTTTTGAC
Phenotype
MIM: 609696 MIM: 606982
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ARID4B PubMed Links

Gene Details

Gene
ARID4B
Gene Name
AT-rich interaction domain 4B
There are no transcripts associated with this gene.

Gene
GGPS1
Gene Name
geranylgeranyl diphosphate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001037277.1 Intron NP_001032354.1

View Full Product Details