Product Details

SNP ID
rs143390167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129171401 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACACAGTTGCATGTGCTCACCT[C/G]TCCAAGCTCTAGAGGGGTATGAAAA
Phenotype
MIM: 116955
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CNBP PubMed Links

Gene Details

Gene
CNBP
Gene Name
CCHC-type zinc finger nucleic acid binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127192.1 Intron NP_001120664.1
NM_001127193.1 Intron NP_001120665.1
NM_001127194.1 Intron NP_001120666.1
NM_001127195.1 Intron NP_001120667.1
NM_001127196.1 Intron NP_001120668.1
NM_003418.4 Intron NP_003409.1

View Full Product Details