Product Details

SNP ID
rs149869494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:47489791 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCAAATGGCAGAGAGGGGTCAGG[A/G]GCCTATGCTGACCTGAGGTCTGGCC
Phenotype
MIM: 615620 MIM: 603215
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KPTN PubMed Links

Gene Details

Gene
KPTN
Gene Name
kaptin, actin binding protein
There are no transcripts associated with this gene.

Gene
NAPA
Gene Name
NSF attachment protein alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003827.3 Intron NP_003818.2
XM_011527436.1 Intron XP_011525738.1
XM_011527437.1 Intron XP_011525739.1
Gene
NAPA-AS1
Gene Name
NAPA antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details