Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001170714.1 | Intron | NP_001164185.1 | |||
NM_001170715.1 | Intron | NP_001164186.1 | |||
NM_001170716.1 | Intron | NP_001164187.1 | |||
NM_001170717.1 | Intron | NP_001164188.1 | |||
NM_001170718.1 | Intron | NP_001164189.1 | |||
NM_001170719.1 | Intron | NP_001164190.1 | |||
NM_001170720.1 | Intron | NP_001164191.1 | |||
NM_001170721.1 | Intron | NP_001164192.1 | |||
NM_014567.3 | Intron | NP_055382.2 | |||
XM_005256258.4 | Intron | XP_005256315.1 | |||
XM_005256259.4 | Intron | XP_005256316.1 | |||
XM_005256260.4 | Intron | XP_005256317.1 | |||
XM_011523475.2 | Intron | XP_011521777.1 | |||
XM_017023897.1 | Intron | XP_016879386.1 | |||
XM_017023898.1 | Intron | XP_016879387.1 | |||
XM_017023899.1 | Intron | XP_016879388.1 |