Product Details

SNP ID
rs10249442
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.7:44801265 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TGCTTGTTTGTGGTTGCCAGTCATA[A/G]TGATTGTTCTTCCTTTTCAAGGTTG
Phenotype
MIM: 123840
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PPIA PubMed Links

Gene Details

Gene
PPIA
Gene Name
peptidylprolyl isomerase A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300981.1 Intron NP_001287910.1
NM_021130.4 Intron NP_066953.1

View Full Product Details