Product Details

SNP ID
hCV25595836
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:41334021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAGTAGGAAATCCCTTGAGCTTA[C/G]ACCTGAGGGAGCGCGCAGTAGCCGG
Phenotype
MIM: 300160
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DDX3X PubMed Links

Gene Details

Gene
DDX3X
Gene Name
DEAD-box helicase 3, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193416.2 714 Intron NP_001180345.1
NM_001193417.2 714 Intron NP_001180346.1
NM_001356.4 714 UTR 5 NP_001347.3
Gene
LOC107985678
Gene Name
uncharacterized LOC107985678
There are no transcripts associated with this gene.

View Full Product Details