Product Details

SNP ID
rs142796743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:75065046 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGAAAGAAGTATACAAGAAGGG[A/G]GCAGGTCTAAAATCTCTGATCACCT
Phenotype
MIM: 300135
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ABCB7 PubMed Links

Gene Details

Gene
ABCB7
Gene Name
ATP binding cassette subfamily B member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271696.1 Intron NP_001258625.1
NM_001271697.1 Intron NP_001258626.1
NM_001271698.1 Intron NP_001258627.1
NM_001271699.1 Intron NP_001258628.1
NM_004299.4 Intron NP_004290.2

View Full Product Details