Product Details

SNP ID
rs12079436
Assay Type
Functionally tested
NCBI dbSNP Submissions
18
Location
Chr.1:3631205 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACCCCCTTGTCCTGCAGGCCAGCAC[A/G]GTGCCTGGAGTGACCCACATAGGCA
Phenotype
MIM: 604266 MIM: 611460 MIM: 606040
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MEGF6 PubMed Links

Gene Details

Gene
MEGF6
Gene Name
multiple EGF like domains 6
There are no transcripts associated with this gene.

Gene
TPRG1L
Gene Name
tumor protein p63 regulated 1-like
There are no transcripts associated with this gene.

Gene
WRAP73
Gene Name
WD repeat containing, antisense to TP73
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017818.3 Intron NP_060288.3
XM_005244754.1 Intron XP_005244811.1
XM_017001387.1 Intron XP_016856876.1

View Full Product Details