Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001134758.3 | 560 | Silent Mutation | NP_001128230.1 | ||
NM_001267544.2 | 560 | UTR 3 | NP_001254473.1 | ||
NM_001267545.2 | 560 | UTR 3 | NP_001254474.1 | ||
NM_001267546.2 | 560 | Silent Mutation | NP_001254475.1 | ||
NM_001267547.2 | 560 | Silent Mutation | NP_001254476.1 | ||
NM_001267548.2 | 560 | Silent Mutation | NP_001254477.1 | ||
NM_001267549.2 | 560 | Silent Mutation | NP_001254478.1 | ||
NM_003224.5 | 560 | Silent Mutation | NP_003215.1 | ||
XM_011528482.2 | 560 | Silent Mutation | XP_011526784.1 | ||
XM_011528483.1 | 560 | Silent Mutation | XP_011526785.1 | ||
XM_017027576.1 | 560 | Silent Mutation | XP_016883065.1 |