Product Details

SNP ID
rs58465018
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.11:67663277 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CGTAACAGCTGCTGGTTCCAGTCGG[C/T]ATAGGGTGGGTAGTGGATCTCCTTT
Phenotype
MIM: 601917
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALDH3B2 PubMed Links

Gene Details

Gene
ALDH3B2
Gene Name
aldehyde dehydrogenase 3 family member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000695.3 1379 Missense Mutation ACC,GCC T366A NP_000686.2
NM_001031615.1 1379 Missense Mutation ACC,GCC T366A NP_001026786.1

View Full Product Details