Product Details

SNP ID
rs114602692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:108742820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAATCCGGGACTCCGAGTCAACT[C/T]TTCATAAAATGTGTGCCAACTTTGC
Phenotype
MIM: 614682
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ETNPPL PubMed Links

Gene Details

Gene
ETNPPL
Gene Name
ethanolamine-phosphate phospho-lyase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146590.1 Intron NP_001140062.1
NM_001146627.1 Intron NP_001140099.1
NM_031279.3 Intron NP_112569.2
XM_006714290.2 Intron XP_006714353.1

View Full Product Details