Product Details

SNP ID
rs149608017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:66554941 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCGGGCCTAAGCATGAGTGTGTGAC[A/G]GAATCGTGATGAGCTAGCAGGAGAT
Phenotype
MIM: 102574 MIM: 603539
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTN3 PubMed Links

Gene Details

Gene
ACTN3
Gene Name
actinin alpha 3 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001104.3 Intron NP_001095.2
NM_001258371.2 Intron NP_001245300.2
Gene
CTSF
Gene Name
cathepsin F
There are no transcripts associated with this gene.

Gene
ZDHHC24
Gene Name
zinc finger DHHC-type containing 24
There are no transcripts associated with this gene.

View Full Product Details