Product Details

SNP ID
rs71449038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:113981629 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCCACCCCCACTGCAATCTCCC[A/G]CCCACTACACTGGCCGTCCAGGGCA
Phenotype
MIM: 605182
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC100507747 PubMed Links

Gene Details

Gene
LOC100507747
Gene Name
uncharacterized LOC100507747
There are no transcripts associated with this gene.

Gene
RASA3
Gene Name
RAS p21 protein activator 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320821.1 Intron NP_001307750.1
NM_001320822.1 Intron NP_001307751.1
NM_007368.3 Intron NP_031394.2
XM_011534841.2 Intron XP_011533143.1
XM_011534842.2 Intron XP_011533144.1
XM_017020438.1 Intron XP_016875927.1
XM_017020439.1 Intron XP_016875928.1

View Full Product Details