Product Details

SNP ID
rs34382193
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:50429232 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACGGTATCTCTCTCCAACTGTGT[C/T]ACATGAATGGAATCTAACTGCTGGC
Phenotype
MIM: 604923
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP4K5 PubMed Links

Gene Details

Gene
MAP4K5
Gene Name
mitogen-activated protein kinase kinase kinase kinase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006575.4 2513 Silent Mutation GTA,GTG V731V NP_006566.2
NM_198794.2 2513 Silent Mutation GTA,GTG V731V NP_942089.1
XM_006720013.3 2513 Silent Mutation GTA,GTG V731V XP_006720076.1
XM_011536377.2 2513 Silent Mutation GTA,GTG V730V XP_011534679.1
XM_011536378.2 2513 Silent Mutation GTA,GTG V664V XP_011534680.1
XM_011536379.2 2513 Silent Mutation GTA,GTG V595V XP_011534681.1
XM_011536380.2 2513 Silent Mutation GTA,GTG V442V XP_011534682.1

View Full Product Details