Product Details

SNP ID
rs571959245
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52420657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCGTTCTTCCCTAGCCATCCTCAT[C/T]CCCTCTCTCCGATCCTTTGCCCTCC
Phenotype
MIM: 608815
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EFHC1 PubMed Links

Gene Details

Gene
EFHC1
Gene Name
EF-hand domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172420.1 Intron NP_001165891.1
NM_018100.3 Intron NP_060570.2

View Full Product Details