Product Details

SNP ID
rs12043303
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:6580423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATATGGCCCCCGGCCCCCGGGA[A/G]GCTGTCAGTGTGTTCCAGCCCTCCG
Phenotype
MIM: 606225 MIM: 165270
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAS1R1 PubMed Links

Gene Details

Gene
TAS1R1
Gene Name
taste 1 receptor member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138697.3 Intron NP_619642.2
NM_177540.2 Intron NP_803884.1
XM_011542203.2 Intron XP_011540505.1
XM_011542206.2 Intron XP_011540508.1
XM_017002402.1 Intron XP_016857891.1
XM_017002403.1 Intron XP_016857892.1
Gene
ZBTB48
Gene Name
zinc finger and BTB domain containing 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278647.1 Intron NP_001265576.1
NM_001278648.1 Intron NP_001265577.1
NM_005341.3 Intron NP_005332.1
XM_017001110.1 Intron XP_016856599.1
XM_017001111.1 Intron XP_016856600.1
XM_017001112.1 Intron XP_016856601.1
XM_017001113.1 Intron XP_016856602.1
XM_017001114.1 Intron XP_016856603.1

View Full Product Details