Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_031475.2 | 748 | Intron | NP_113663.2 | ||
XM_005263501.2 | 748 | Intron | XP_005263558.1 | ||
XM_011542231.1 | 748 | Intron | XP_011540533.1 | ||
XM_011542232.1 | 748 | Intron | XP_011540534.1 | ||
XM_011542233.2 | 748 | Intron | XP_011540535.1 | ||
XM_011542236.2 | 748 | Intron | XP_011540538.1 | ||
XM_011542237.1 | 748 | Intron | XP_011540539.1 | ||
XM_011542238.2 | 748 | Intron | XP_011540540.1 | ||
XM_017002433.1 | 748 | Intron | XP_016857922.1 | ||
XM_017002434.1 | 748 | Intron | XP_016857923.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039664.1 | 748 | Intron | NP_001034753.1 | ||
NM_003790.2 | 748 | Silent Mutation | NP_003781.1 | ||
NM_148965.1 | 748 | Silent Mutation | NP_683866.1 | ||
NM_148966.1 | 748 | Silent Mutation | NP_683867.1 | ||
NM_148967.1 | 748 | Silent Mutation | NP_683868.1 | ||
NM_148970.1 | 748 | Silent Mutation | NP_683871.1 |