Product Details

SNP ID
rs138678761
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:80082210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATGCCATAGTATTGTGCAAGTAAG[G/T]CTTTGAAGTAGGTGTTTGTTGCCTT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM254 PubMed Links

Gene Details

Gene
TMEM254
Gene Name
transmembrane protein 254
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270367.1 311 Intron NP_001257296.1
NM_001270368.1 311 UTR 3 NP_001257297.1
NM_001270369.1 311 Intron NP_001257298.1
NM_001270370.1 311 Intron NP_001257299.1
NM_001270371.1 311 Intron NP_001257300.1
NM_001270372.1 311 UTR 3 NP_001257301.1
NM_001270373.1 311 Intron NP_001257302.1
NM_001270374.1 311 Intron NP_001257303.1
NM_025125.3 311 Intron NP_079401.2
XM_006717978.2 311 Intron XP_006718041.1
XM_011540192.2 311 UTR 3 XP_011538494.1
XM_011540193.2 311 Intron XP_011538495.1
XM_017016679.1 311 Intron XP_016872168.1
XM_017016680.1 311 Intron XP_016872169.1
Gene
TMEM254-AS1
Gene Name
TMEM254 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details