Product Details
- SNP ID
-
rs61735589
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:49280926 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCGGCACGAGGCGTCGGCTGACCTG[G/T]ATGGTTTGTCTGTTGTAGACTTTTA
- Phenotype
-
MIM: 600432
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CBLN1
PubMed Links
Gene Details
- Gene
- CBLN1
- Gene Name
- cerebellin 1 precursor
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004352.3 |
747 |
Silent Mutation |
ATA,ATC |
I127I |
NP_004343.1 |
View Full Product Details