Product Details

SNP ID
hCV25599581
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:72097749 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTAGGGTACTGTATAAGCCGCAGT[C/G]AGGCTGGGGGCCAAGGGGGTGGGGT
Phenotype
MIM: 614717 MIM: 613510 MIM: 612414
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ANAPC15 PubMed Links

Gene Details

Gene
ANAPC15
Gene Name
anaphase promoting complex subunit 15
There are no transcripts associated with this gene.

Gene
LAMTOR1
Gene Name
late endosomal/lysosomal adaptor, MAPK and MTOR activator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017907.2 722 UTR 3 NP_060377.1
Gene
LRTOMT
Gene Name
leucine rich transmembrane and O-methyltransferase domain containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145307.4 722 Intron NP_001138779.1
NM_001145308.4 722 Intron NP_001138780.1
NM_001145309.3 722 Intron NP_001138781.1
NM_001145310.3 722 Intron NP_001138782.1
NM_001205138.3 722 Intron NP_001192067.1
NM_001271471.2 722 Intron NP_001258400.1
NM_001318803.1 722 Intron NP_001305732.1
NM_145309.5 722 Intron NP_660352.1

View Full Product Details