Product Details

SNP ID
rs191329339
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:67687486 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTTTAATAGGTAATTACCCTTAA[C/T]GCTTTTGCAGGGAACCCAACTACCT
Phenotype
MIM: 126335
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GADD45A PubMed Links

Gene Details

Gene
GADD45A
Gene Name
growth arrest and DNA damage inducible alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199741.1 Intron NP_001186670.1
NM_001199742.1 Intron NP_001186671.1
NM_001924.3 Intron NP_001915.1

View Full Product Details