Product Details

SNP ID
rs3093423
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:206899627 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGTCCTTCACAGCTTGCTAATC[A/C]GTTTCCAGTTTCCCATATAATAACT
Phenotype
MIM: 606015 MIM: 604136
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FCMR PubMed Links

Gene Details

Gene
FCMR
Gene Name
Fc fragment of IgM receptor
There are no transcripts associated with this gene.

Gene
IL24
Gene Name
interleukin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001185156.1 Intron NP_001172085.1
NM_001185157.1 Intron NP_001172086.1
NM_001185158.1 Intron NP_001172087.1
NM_006850.3 Intron NP_006841.1
XM_011509101.2 Intron XP_011507403.1
XM_017000121.1 Intron XP_016855610.1

View Full Product Details