Product Details

SNP ID
rs34841326
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:191836575 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTCCAGGGCCTCCTCATCGTGG[G/T]GCAAATCATCATCTGAGGAGAGGTC
Phenotype
MIM: 606728
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SDPR PubMed Links

Gene Details

Gene
SDPR
Gene Name
serum deprivation response
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004657.5 981 Missense Mutation NP_004648.1

View Full Product Details