Product Details

SNP ID
rs200966022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64789814 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAGACAGGTGGGAAGCACTGAGG[C/T]CACTCCAGTAGGTCCTTTCCAAAGA
Phenotype
MIM: 603166
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP4K2 PubMed Links

Gene Details

Gene
MAP4K2
Gene Name
mitogen-activated protein kinase kinase kinase kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307990.1 Intron NP_001294919.1
NM_004579.4 Intron NP_004570.2
XM_011545203.2 Intron XP_011543505.1
XM_011545204.2 Intron XP_011543506.1
XM_017018093.1 Intron XP_016873582.1
XM_017018094.1 Intron XP_016873583.1
XM_017018095.1 Intron XP_016873584.1
XM_017018096.1 Intron XP_016873585.1

View Full Product Details