Product Details
- SNP ID
-
hCV25601097
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:67399702 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACTAAGTTTTTGTAGTTTCTTTAAA[A/T]GAAAAGTATTGTCTATTATAATTCT
- Phenotype
-
MIM: 613196
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ETAA1
PubMed Links
Gene Details
- Gene
- ETAA1
- Gene Name
- Ewing tumor associated antigen 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_019002.3 |
|
Intron |
|
|
NP_061875.2 |
View Full Product Details