Product Details

SNP ID
rs16859723
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:182842487 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAGATTCAGTAATCTAGATAATTG[C/G]TTCCTCCCAGTTTTTGCTATTATAA
Phenotype
MIM: 603115
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DHX9 PubMed Links

Gene Details

Gene
DHX9
Gene Name
DEAH-box helicase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001357.4 Intron NP_001348.2
Gene
LOC647070
Gene Name
nascent polypeptide-associated complex subunit alpha, muscle-specific form-like
There are no transcripts associated with this gene.

View Full Product Details