Product Details

SNP ID
rs73530370
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3936554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTGGGGGGAGCCCAGGCAGTCTC[A/G]TGCACACGCTGTCTCCCCAGCATGA
Phenotype
MIM: 608179 MIM: 608705
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ATCAY PubMed Links

Gene Details

Gene
ATCAY
Gene Name
ATCAY, caytaxin
There are no transcripts associated with this gene.

Gene
NMRK2
Gene Name
nicotinamide riboside kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289117.1 Intron NP_001276046.1
NM_170678.2 Intron NP_733778.1
XM_006722725.3 Intron XP_006722788.1
XM_006722726.3 Intron XP_006722789.1
XM_017026597.1 Intron XP_016882086.1

View Full Product Details