Product Details

SNP ID
rs80106507
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:8682782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCGATACCTCGCGAGACTTGGC[A/G]AAGGCCTTCCTTTTTCGTCTGGGCT
Phenotype
MIM: 603637 MIM: 611333 MIM: 612000
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPL27A PubMed Links

Gene Details

Gene
RPL27A
Gene Name
ribosomal protein L27a
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000990.4 436 UTR 5 NP_000981.1
Gene
SNORA3A
Gene Name
small nucleolar RNA, H/ACA box 3A
There are no transcripts associated with this gene.

Gene
SNORA3B
Gene Name
small nucleolar RNA, H/ACA box 3B
There are no transcripts associated with this gene.

Gene
TRIM66
Gene Name
tripartite motif containing 66
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014818.1 436 Intron NP_055633.1
XM_006718397.2 436 Intron XP_006718460.1
XM_006718398.3 436 Intron XP_006718461.1
XM_011520504.1 436 Intron XP_011518806.1
XM_011520507.2 436 Intron XP_011518809.1
XM_011520508.1 436 Intron XP_011518810.1
XM_011520509.1 436 Intron XP_011518811.1
XM_011520510.1 436 Intron XP_011518812.1
XM_011520511.1 436 Intron XP_011518813.1
XM_011520512.1 436 Intron XP_011518814.1
XM_011520513.1 436 Intron XP_011518815.1
XM_011520514.2 436 Intron XP_011518816.1
XM_011520515.1 436 Intron XP_011518817.1
XM_011520516.1 436 Intron XP_011518818.1
XM_011520517.2 436 Intron XP_011518819.1
XM_011520518.1 436 Intron XP_011518820.1
XM_011520519.1 436 Intron XP_011518821.1
XM_011520522.2 436 Intron XP_011518824.1
XM_011520523.2 436 Intron XP_011518825.1
XM_011520524.1 436 Intron XP_011518826.1
XM_011520525.1 436 Intron XP_011518827.1
XM_011520526.2 436 Intron XP_011518828.1
XM_011520527.1 436 Intron XP_011518829.1
XM_017018629.1 436 UTR 5 XP_016874118.1
XM_017018630.1 436 Intron XP_016874119.1
XM_017018631.1 436 Intron XP_016874120.1

View Full Product Details