Product Details

SNP ID
rs769538147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8321580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCAAGCTGGCCAGGGTGAGGTGGG[A/G]GCCCGAATTTGGGGGCAGGGGGAGG
Phenotype
MIM: 614611 MIM: 602139 MIM: 603685
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KANK3 PubMed Links

Gene Details

Gene
KANK3
Gene Name
KN motif and ankyrin repeat domains 3
There are no transcripts associated with this gene.

Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005001.4 Intron NP_004992.2
Gene
RPS28
Gene Name
ribosomal protein S28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031.4 Intron NP_001022.1

View Full Product Details