Product Details

SNP ID
rs142443316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:67202668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGAAAAAAAAGATTTCTCCTTAA[C/T]CTCCTACTAAAAACTAAAATTAAGG
Phenotype
MIM: 614888 MIM: 603109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AAGAB PubMed Links

Gene Details

Gene
AAGAB
Gene Name
alpha- and gamma-adaptin binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271885.1 1456 UTR 3 NP_001258814.1
NM_001271886.1 1456 UTR 3 NP_001258815.1
NM_024666.4 1456 UTR 3 NP_078942.3
XM_011522020.1 1456 UTR 3 XP_011520322.1
Gene
SMAD3
Gene Name
SMAD family member 3
There are no transcripts associated with this gene.

View Full Product Details