Product Details

SNP ID
hCV25602561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144424671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTACGCTCGTGCCCAGCTGCAGCA[A/G]GCAGCCTCGGCTCTCCTAACCACGT
Phenotype
MIM: 607059 MIM: 604546 MIM: 611952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC39A4 PubMed Links

Gene Details

Gene
SLC39A4
Gene Name
solute carrier family 39 member 4
There are no transcripts associated with this gene.

Gene
TONSL
Gene Name
tonsoku-like, DNA repair protein
There are no transcripts associated with this gene.

Gene
VPS28
Gene Name
VPS28, ESCRT-I subunit
There are no transcripts associated with this gene.

View Full Product Details