Product Details

SNP ID
hCV25602626
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:44970130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCACTGGCCGGCACTCCTCTGG[A/C]TCACCTGTCTGTCCACCCTTCCTTG
Phenotype
MIM: 611309 MIM: 610559
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C10orf10 PubMed Links

Gene Details

Gene
C10orf10
Gene Name
chromosome 10 open reading frame 10
There are no transcripts associated with this gene.

Gene
RASSF4
Gene Name
Ras association domain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032023.3 Intron NP_114412.2

View Full Product Details