Product Details

SNP ID
rs145465075
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6482197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAGGCGTTGCTGCTGAACAGCC[C/T]GGGGTCTCTCCATCAGGCAGCTAGC
Phenotype
MIM: 601638 MIM: 607388 MIM: 605493
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ARFIP2 PubMed Links
Additional Information
For this assay, SNP(s) [rs116095394] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
ARFIP2
Gene Name
ADP ribosylation factor interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242854.1 358 Intron NP_001229783.1
NM_001242855.1 358 Intron NP_001229784.1
NM_001242856.1 358 Intron NP_001229785.1
NM_012402.3 358 Intron NP_036534.1
XM_005252840.4 358 Intron XP_005252897.1
Gene
TIMM10B
Gene Name
translocase of inner mitochondrial membrane 10 homolog B (yeast)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012192.3 358 Silent Mutation CCC,CCT P96P NP_036324.1
Gene
TRIM3
Gene Name
tripartite motif containing 3
There are no transcripts associated with this gene.

View Full Product Details