Product Details

SNP ID
rs115801368
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:6401146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAGCCCCACCAGCAGGGCATAAG[C/T]TGGACACTTGCCCAGCCGAGGCCCT
Phenotype
MIM: 602709 MIM: 607608
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APBB1 PubMed Links

Gene Details

Gene
APBB1
Gene Name
amyloid beta precursor protein binding family B member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164.4 Intron NP_001155.1
NM_001257319.2 Intron NP_001244248.1
NM_001257320.2 Intron NP_001244249.1
NM_001257321.2 Intron NP_001244250.1
NM_001257323.2 Intron NP_001244252.1
NM_001257325.2 Intron NP_001244254.1
NM_001257326.2 Intron NP_001244255.1
NM_145689.2 Intron NP_663722.1
XM_006718209.3 Intron XP_006718272.2
XM_006718210.3 Intron XP_006718273.2
XM_006718211.1 Intron XP_006718274.1
XM_006718212.3 Intron XP_006718275.2
XM_006718213.1 Intron XP_006718276.1
XM_006718214.3 Intron XP_006718277.2
XM_006718215.3 Intron XP_006718278.3
XM_011520037.2 Intron XP_011518339.1
XM_011520038.2 Intron XP_011518340.1
XM_011520040.2 Intron XP_011518342.1
XM_011520042.1 Intron XP_011518344.1
XM_017017639.1 Intron XP_016873128.1
XM_017017640.1 Intron XP_016873129.1
XM_017017641.1 Intron XP_016873130.1
XM_017017642.1 Intron XP_016873131.1
Gene
SMPD1
Gene Name
sphingomyelin phosphodiesterase 1
There are no transcripts associated with this gene.

View Full Product Details