Product Details

SNP ID
rs61731123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:3491774 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGCAGCTGTCCCAGGACGTAGCC[A/G]ACCTCGTGGCGGAAGAGGGCGCTCC
Phenotype
MIM: 611262
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DOHH PubMed Links

Gene Details

Gene
DOHH
Gene Name
deoxyhypusine hydroxylase/monooxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145165.1 790 Silent Mutation GTC,GTT V209V NP_001138637.1
NM_031304.4 790 Silent Mutation GTC,GTT V209V NP_112594.1
XM_011528341.2 790 Intron XP_011526643.1

View Full Product Details