Product Details
- SNP ID
-
rs7157941
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:102326152 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATCGCCAGCCCTCAGCTGCTCAG[A/G]GTACAGCCGCTTGTGAGAACCGAGC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF839
PubMed Links
Gene Details
- Gene
- ZNF839
- Gene Name
- zinc finger protein 839
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001267827.1 |
471 |
Silent Mutation |
AGA,AGG |
R36R |
NP_001254756.1 |
NM_001267828.1 |
471 |
Silent Mutation |
AGA,AGG |
R36R |
NP_001254757.1 |
NM_018335.4 |
471 |
Silent Mutation |
AGA,AGG |
R152R |
NP_060805.3 |
XM_006720203.3 |
471 |
Silent Mutation |
AGA,AGG |
R64R |
XP_006720266.2 |
XM_011536945.1 |
471 |
Silent Mutation |
AGA,AGG |
R186R |
XP_011535247.1 |
XM_011536946.2 |
471 |
Silent Mutation |
AGA,AGG |
R186R |
XP_011535248.1 |
XM_011536948.2 |
471 |
Silent Mutation |
AGA,AGG |
R36R |
XP_011535250.1 |
XM_011536949.2 |
471 |
Silent Mutation |
AGA,AGG |
R186R |
XP_011535251.1 |
XM_011536950.2 |
471 |
Silent Mutation |
AGA,AGG |
R186R |
XP_011535252.1 |
XM_017021447.1 |
471 |
Silent Mutation |
AGA,AGG |
R152R |
XP_016876936.1 |
XM_017021448.1 |
471 |
Silent Mutation |
AGA,AGG |
R152R |
XP_016876937.1 |
XM_017021449.1 |
471 |
Intron |
|
|
XP_016876938.1 |
XM_017021450.1 |
471 |
Silent Mutation |
AGA,AGG |
R82R |
XP_016876939.1 |
XM_017021451.1 |
471 |
Silent Mutation |
AGA,AGG |
R36R |
XP_016876940.1 |
View Full Product Details