Product Details

SNP ID
rs34540052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:6445125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACGGCCACATCCCTGGTGGCTGTG[C/T]GTTCTGGGGACCCTGGTGGGGCTCT
Phenotype
MIM: 186711 MIM: 607081
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD27 PubMed Links

Gene Details

Gene
CD27
Gene Name
CD27 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001242.4 259 Silent Mutation TGC,TGT C10C NP_001233.1
XM_011521042.2 259 Intron XP_011519344.1
XM_017020232.1 259 Silent Mutation TGC,TGT C10C XP_016875721.1
XM_017020233.1 259 Intron XP_016875722.1
XM_017020234.1 259 Silent Mutation TGC,TGT C10C XP_016875723.1
Gene
CD27-AS1
Gene Name
CD27 antisense RNA 1
There are no transcripts associated with this gene.

Gene
TAPBPL
Gene Name
TAP binding protein like
There are no transcripts associated with this gene.

View Full Product Details