Product Details

SNP ID
rs3768235
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:85267691 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
CATCTCACTAGAGTTTGCACAAGTT[C/T]CTTCTTCTTCTAACTGTAGATCTGG
Phenotype
MIM: 603517
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCL10 PubMed Links

Gene Details

Gene
BCL10
Gene Name
B-cell CLL/lymphoma 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320715.1 1059 Missense Mutation GAA,GGA E202G NP_001307644.1
NM_003921.4 1059 Missense Mutation GAA,GGA E213G NP_003912.1
XM_011542397.2 1059 Missense Mutation GAA,GGA E266G XP_011540699.1
XM_011542398.2 1059 Missense Mutation GAA,GGA E255G XP_011540700.1
XM_011542399.2 1059 Missense Mutation GAA,GGA E195G XP_011540701.1
Gene
C1orf52
Gene Name
chromosome 1 open reading frame 52
There are no transcripts associated with this gene.

Gene
LOC646626
Gene Name
uncharacterized LOC646626
There are no transcripts associated with this gene.

View Full Product Details