Product Details

SNP ID
rs74058358
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:16978148 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGCTGAGTTCTGGTCATAAGTCG[C/T]GACAAAGGCTCACTATGAATTCCCC
Phenotype
MIM: 610513 MIM: 615776 MIM: 156790
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ATP13A2 PubMed Links

Gene Details

Gene
ATP13A2
Gene Name
ATPase 13A2
There are no transcripts associated with this gene.

Gene
CROCC
Gene Name
ciliary rootlet coiled-coil, rootletin
There are no transcripts associated with this gene.

Gene
MFAP2
Gene Name
microfibrillar associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135247.1 Intron NP_001128719.1
NM_001135248.1 Intron NP_001128720.1
NM_002403.3 Intron NP_002394.1
NM_017459.2 Intron NP_059453.1
XM_011541476.2 Intron XP_011539778.1

View Full Product Details