Product Details
- SNP ID
-
rs34725707
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:68684970 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGCTCGCCTCCCTCCTCCTCGCCG[C/T]GGCCCTTTCTGCCTCTGCGGGGCTC
- Phenotype
-
MIM: 137035
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GAL
PubMed Links
Gene Details
- Gene
- GAL
- Gene Name
- galanin and GMAP prepropeptide
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_015973.4 |
305 |
Missense Mutation |
GCG,GTG |
A16V |
NP_057057.2 |
View Full Product Details