Product Details
- SNP ID
-
rs35537366
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:6749867 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCACTCTCATCCAGGTTGATATA[A/G]TCCTGCCGCTCCTCCTGGTCCCCCG
- Phenotype
-
MIM: 601401
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MLF2
PubMed Links
Gene Details
- Gene
- MLF2
- Gene Name
- myeloid leukemia factor 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005439.2 |
603 |
Silent Mutation |
GAC,GAT |
D180D |
NP_005430.1 |
View Full Product Details