Product Details

SNP ID
rs35249920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:121024015 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTCACGTTCCAGCTGGAGATGGGG[A/T]TCTCCCTGTTGTCATAGCAACAGTC
Phenotype
MIM: 603281
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C12orf43 PubMed Links

Gene Details

Gene
C12orf43
Gene Name
chromosome 12 open reading frame 43
There are no transcripts associated with this gene.

Gene
OASL
Gene Name
2'-5'-oligoadenylate synthetase like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261825.1 1293 Intron NP_001248754.1
NM_003733.3 1293 Missense Mutation AAC,ATC N341I NP_003724.1
NM_198213.2 1293 UTR 3 NP_937856.1
XM_017020140.1 1293 Missense Mutation AAC,ATC N341I XP_016875629.1
XM_017020141.1 1293 UTR 3 XP_016875630.1

View Full Product Details