Product Details

SNP ID
rs41272777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47973271 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAGAAAGAGAGGGGAGAAAAGTCC[A/G]AACTGTGAGAGGGTGGGATGAATGG
Phenotype
MIM: 120140
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COL2A1 PubMed Links

Gene Details

Gene
COL2A1
Gene Name
collagen type II alpha 1 chain
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001844.4 6166 UTR 3 NP_001835.3
NM_033150.2 6166 UTR 3 NP_149162.2
XM_017018828.1 6166 UTR 3 XP_016874317.1
XM_017018829.1 6166 UTR 3 XP_016874318.1
XM_017018830.1 6166 UTR 3 XP_016874319.1
XM_017018831.1 6166 UTR 3 XP_016874320.1
Gene
TMEM106C
Gene Name
transmembrane protein 106C
There are no transcripts associated with this gene.

View Full Product Details