Product Details

SNP ID
rs35500826
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47965905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTGGTTTGGTGGTTTTCTTCCTG[C/T]TTCCGCATTCAGTCCTTGTGGATGA
Phenotype
MIM: 120140
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
COL2A1 PubMed Links

Gene Details

Gene
COL2A1
Gene Name
collagen type II alpha 1 chain
There are no transcripts associated with this gene.

Gene
TMEM106C
Gene Name
transmembrane protein 106C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143841.1 505 Missense Mutation CTT,TTT L107F NP_001137313.1
NM_001143842.1 505 Missense Mutation CTT,TTT L107F NP_001137314.1
NM_001143843.1 505 Missense Mutation CTT,TTT L107F NP_001137315.1
NM_024056.3 505 Missense Mutation CTT,TTT L107F NP_076961.1

View Full Product Details