Product Details
- SNP ID
-
rs73996007
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75277224 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCACATGCTTAGGGATCTCAAGG[A/G]ATGGACGCAGGTGAAGGGGCCATGG
- Phenotype
-
MIM: 612072
MIM: 606521
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
LOC100287042
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs192824470] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LOC100287042
- Gene Name
- uncharacterized LOC100287042
There are no transcripts associated with this gene.
- Gene
- MIF4GD
- Gene Name
- MIF4G domain containing
There are no transcripts associated with this gene.
- Gene
- SLC25A19
- Gene Name
- solute carrier family 25 member 19
View Full Product Details