Product Details
- SNP ID
-
hCV25607724
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:120214006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCGTGGAACCCTGGGGAGCGGGGGT[C/T]TTGGAGGCACAGTTCATTCCGGCTT
- Phenotype
-
MIM: 602505
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PXN
PubMed Links
Gene Details
- Gene
- PXN
- Gene Name
- paxillin
- Gene
- PXN-AS1
- Gene Name
- PXN antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details