Product Details

SNP ID
rs113762053
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:105855988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGTTCGTCTGTGCCCTGCATGAC[C/G]TCCTTGGAAGGCAGCAGCACCTGTG
Phenotype
MIM: 146910 MIM: 147070 MIM: 147010 MIM: 147020
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IGH PubMed Links

Gene Details

Gene
IGH
Gene Name
immunoglobulin heavy locus
There are no transcripts associated with this gene.

Gene
IGHM
Gene Name
immunoglobulin heavy constant mu
There are no transcripts associated with this gene.

Gene
MIR4507
Gene Name
microRNA 4507
There are no transcripts associated with this gene.

Gene
MIR4537
Gene Name
microRNA 4537
There are no transcripts associated with this gene.

Gene
MIR4538
Gene Name
microRNA 4538
There are no transcripts associated with this gene.

Gene
MIR4539
Gene Name
microRNA 4539
There are no transcripts associated with this gene.

View Full Product Details