Product Details

SNP ID
rs3780915
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:14521098 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCAAGTCAAAATTCTCTGCTGGCTG[A/G]AATATTCTCCAGGGGCTTTTGCCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM107B PubMed Links

Gene Details

Gene
FAM107B
Gene Name
family with sequence similarity 107 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282695.1 644 UTR 3 NP_001269624.1
NM_001282696.1 644 UTR 3 NP_001269625.1
NM_001282697.1 644 UTR 3 NP_001269626.1
NM_001282698.1 644 UTR 3 NP_001269627.1
NM_001282699.1 644 UTR 3 NP_001269628.1
NM_001282700.1 644 UTR 3 NP_001269629.1
NM_001282701.1 644 UTR 3 NP_001269630.1
NM_001282702.1 644 UTR 3 NP_001269631.1
NM_001282703.1 644 UTR 3 NP_001269632.1
NM_001320735.1 644 UTR 3 NP_001307664.1
NM_001320736.1 644 UTR 3 NP_001307665.1
NM_001320737.1 644 UTR 3 NP_001307666.1
NM_001320738.1 644 UTR 3 NP_001307667.1
NM_001320739.1 644 UTR 3 NP_001307668.1
NM_001320740.1 644 UTR 3 NP_001307669.1
NM_001320741.1 644 UTR 3 NP_001307670.1
NM_031453.3 644 UTR 3 NP_113641.2
XM_005252616.3 644 UTR 3 XP_005252673.1
XM_011519735.1 644 UTR 3 XP_011518037.1
XM_017016746.1 644 UTR 3 XP_016872235.1
XM_017016747.1 644 UTR 3 XP_016872236.1
XM_017016748.1 644 UTR 3 XP_016872237.1
XM_017016749.1 644 UTR 3 XP_016872238.1

View Full Product Details